Anti-SRM Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SRM
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77857-954
- 77740-777
- Antigen Symbol:
- SRM
- SRM
- Antigen Synonyms:
- OTTHUMP00000002170|PAPT|Putrescine aminopropyltransferase|SPDSY|SPEE_HUMAN|Spermidine synthase 1|Spermidine synthase|SPS1|Srm|SRML1
- OTTHUMP00000002170|PAPT|Putrescine aminopropyltransferase|SPDSY|SPEE_HUMAN|Spermidine synthase 1|Spermidine synthase|SPS1|Srm|SRML1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P19623
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Spermidine synthase protein (17-302AA)
- Recombinant Human Spermidine synthase protein (17-302AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice