Anti-MEMO1 Rabbit Polyclonal Antibody
Catalog # 77743-001
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MEMO1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9Y316
- Antigen Synonyms:C21orf19-like protein|C2orf4|CGI 27|DKFZp434I0135|FLJ25031|HCV NS5A-transactivated protein 7|Hepatitis C virus NS5A-transactivated protein 7|Mediator of cell motility 1|Mediator of ErbB2-driven cell motility 1|MEMO|Memo-1|MEMO1_HUMAN|NS5ATP7|Protein MEMO1
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Protein MEMO1
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MEMO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat