Anti-NUCB2 Rabbit Recombinant polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NUCB2
Clonality: Recombinant polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77745-138
- 77745-139
- Antigen Symbol:
- NUCB2
- NUCB2
- Antigen Synonyms:
- DNA binding protein NEFA| DNA-binding protein NEFA| Gastric cancer antigen Zg4| HEL S 109| NEFA| Nefastin 1| Nefastin-1| Nefastin1| NUCB2_HUMAN| Nucleobindin 2| Nucleobindin-2| Nucleobinding 2| Prepronefastin| Prepronesfatin
- DNA binding protein NEFA| DNA-binding protein NEFA| Gastric cancer antigen Zg4| HEL S 109| NEFA| Nefastin 1| Nefastin-1| Nefastin1| NUCB2_HUMAN| Nucleobindin 2| Nucleobindin-2| Nucleobinding 2| Prepronefastin| Prepronesfatin
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P80303
- P80303
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Nucleobindin-2 protein (25-106AA)
- Nucleobindin-2 protein (25-106AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice