Anti-ITGA2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ITGA2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77741-258
- 77858-257
- Antigen Symbol:
- ITGA2
- ITGA2
- Antigen Synonyms:
- BR|CD 49b|CD49 antigen like family member B|CD49 antigen-like family member B|CD49b|CD49b antigen|Collagen receptor|DX5|Glycoprotein Ia deficiency included|GP Ia|GP Ia deficiency|included|GPIa|HPA 5 included|HPA5 included|Human platelet alloantigen system 5|Integrin alpha 2|Integrin alpha II|Integrin alpha-2|Integrin|alpha 2 (CD49B alpha 2 subunit of VLA 2 receptor)|ITA2_HUMAN|Platelet alloantigen Br(a)|Platelet antigen Br|Platelet glycoprotein GPIa|Platelet glycoprotein Ia|Platelet glycoprotein Ia/IIa|Platelet membrane glycoprotein Ia|Platelet receptor for collagen|deficiency of|Very late activation protein 2 receptor alpha 2 subunit|VLA 2 alpha chain|VLA 2|VLA 2 subunit alpha|VLA-2 subunit alpha|VLA2|VLA2 receptor alpha 2 subunit|VLAA2
- BR|CD 49b|CD49 antigen like family member B|CD49 antigen-like family member B|CD49b|CD49b antigen|Collagen receptor|DX5|Glycoprotein Ia deficiency included|GP Ia|GP Ia deficiency|included|GPIa|HPA 5 included|HPA5 included|Human platelet alloantigen system 5|Integrin alpha 2|Integrin alpha II|Integrin alpha-2|Integrin|alpha 2 (CD49B alpha 2 subunit of VLA 2 receptor)|ITA2_HUMAN|Platelet alloantigen Br(a)|Platelet antigen Br|Platelet glycoprotein GPIa|Platelet glycoprotein Ia|Platelet glycoprotein Ia/IIa|Platelet membrane glycoprotein Ia|Platelet receptor for collagen|deficiency of|Very late activation protein 2 receptor alpha 2 subunit|VLA 2 alpha chain|VLA 2|VLA 2 subunit alpha|VLA-2 subunit alpha|VLA2|VLA2 receptor alpha 2 subunit|VLAA2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P17301
- P17301
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Integrin alpha-2 protein (188-365AA)
- Recombinant Human Integrin alpha-2 protein (188-365AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together