Anti-NME4 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: NME4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-548
- 77744-549
- Antigen Symbol:
- NME4
- NME4
- Antigen Synonyms:
- metastatic inhibition factor NM23H4|mitochondrial|NDK|NDKM_HUMAN|NDP kinase|NDP kinase D|NDP kinase, mitochondrial|NDPK D|NDPKD|nm23 H4|nm23-H4|NM23D|NM23H4|Nm23M4|NME/NM23 nucleoside diphosphate kinase 4|NME4|Non metastatic cells 4 protein expressed in|Non metastatic protein 23, homolog 4|Nucleoside diphosphate kinase D|Nucleoside diphosphate kinase, mitochondrial
- metastatic inhibition factor NM23H4|mitochondrial|NDK|NDKM_HUMAN|NDP kinase|NDP kinase D|NDP kinase, mitochondrial|NDPK D|NDPKD|nm23 H4|nm23-H4|NM23D|NM23H4|Nm23M4|NME/NM23 nucleoside diphosphate kinase 4|NME4|Non metastatic cells 4 protein expressed in|Non metastatic protein 23, homolog 4|Nucleoside diphosphate kinase D|Nucleoside diphosphate kinase, mitochondrial
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00746
- O00746
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Nucleoside diphosphate kinase, mitochondrial protein (1-187AA)
- Nucleoside diphosphate kinase, mitochondrial protein (1-187AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice