Anti-NDUFB3 Rabbit Recombinant polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NDUFB3
Clonality: Recombinant polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-175
- 77744-176
- Antigen Symbol:
- NDUFB3
- NDUFB3
- Antigen Synonyms:
- B12| CI B12| CI-B12| Complex I B12| Complex I-B12| NADH dehydrogenase (ubiquinone) 1 beta subcomplex 3 12kDa| NADH dehydrogenase (ubiquinone) 1 beta subcomplex 3| NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 3| NADH ubiquinone oxidoreductase B12 subunit| NADH-ubiquinone oxidoreductase 1 beta subcomplex| 3| NADH-ubiquinone oxidoreductase B12 subunit| NADH:ubiquinone oxidoreductase subunit B3| NDUB3_HUMAN| NDUFB 3| NDUFB3
- B12| CI B12| CI-B12| Complex I B12| Complex I-B12| NADH dehydrogenase (ubiquinone) 1 beta subcomplex 3 12kDa| NADH dehydrogenase (ubiquinone) 1 beta subcomplex 3| NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 3| NADH ubiquinone oxidoreductase B12 subunit| NADH-ubiquinone oxidoreductase 1 beta subcomplex| 3| NADH-ubiquinone oxidoreductase B12 subunit| NADH:ubiquinone oxidoreductase subunit B3| NDUB3_HUMAN| NDUFB 3| NDUFB3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43676
- O43676
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 3 protein (2-65AA)
- NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 3 protein (2-65AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice