Anti-HEXIM1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: HEXIM1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77739-656
- 77856-833
- Antigen Symbol:
- HEXIM1
- HEXIM1
- Antigen Synonyms:
- Cardiac lineage protein 1|CLP 1|CLP1|EDG 1|EDG1|Estrogen down-regulated gene 1 protein|FLJ13562|Hexamethylene bis acetamide inducible 1|Hexamethylene bis acetamide inducible protein|Hexamethylene bis acetamide inducible transcript 1|Hexamethylene bis-acetamide-inducible protein 1|HEXI1_HUMAN|HEXIM 1|HEXIM1 protein|HIS 1|HIS1|HMBA inducible|MAQ 1|MAQ1|Menage a quatre 1|Menage a quatre protein 1|Protein HEXIM1
- Cardiac lineage protein 1|CLP 1|CLP1|EDG 1|EDG1|Estrogen down-regulated gene 1 protein|FLJ13562|Hexamethylene bis acetamide inducible 1|Hexamethylene bis acetamide inducible protein|Hexamethylene bis acetamide inducible transcript 1|Hexamethylene bis-acetamide-inducible protein 1|HEXI1_HUMAN|HEXIM 1|HEXIM1 protein|HIS 1|HIS1|HMBA inducible|MAQ 1|MAQ1|Menage a quatre 1|Menage a quatre protein 1|Protein HEXIM1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O94992
- O94992
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Protein HEXIM1 protein (210-359AA)
- Recombinant Human Protein HEXIM1 protein (210-359AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice