Anti-NBL1 Rabbit Polyclonal Antibody
77743-966
New Product
- Antigen Symbol:NBL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P41271
- Antigen Synonyms:D1S1733E|DAN|DAN domain family member 1|DAND 1|DAND1|Differential screening selected gene aberrant in neuroblastoma|MGC8972|NB|NBL 1|NBL1_HUMAN|Neuroblastoma candidate region suppression of tumorigenicity 1|Neuroblastoma suppression of tumorigenicity 1|Neuroblastoma suppressor of tumorigenicity 1|NO 3|NO3|Protein N03|Zinc finger protein DAN
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Zinc finger protein DAN
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: NBL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat