Anti-HK3 Rabbit Polyclonal Antibody
Catalog # 77740-102
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HK3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Form:Liquid
- Gene ID:P52790
- Antigen Synonyms:ATP:D hexose 6 phosphotransferase|Hexokinase type III|Hexokinase-3|Hexokinase3|HK III|Hk3|HKIII|HXK3|HXK3_HUMAN
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:HXK3_HUMAN
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: HK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat