Anti-LDHC Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LDHC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-880
- 77742-047
- Antigen Symbol:
- LDHC
- LDHC
- Antigen Synonyms:
- Cancer/testis antigen 32| CT32| EC 1.1.1.27| L lactate dehydrogenase C chain| L-lactate dehydrogenase C chain| Lactate Dehydrogenase C| Lactate dehydrogenase c variant 1| Lactate dehydrogenase c variant 3| Lactate dehydrogenase c variant 4| Lactate dehydrogenase C4| LDH C| LDH testis subunit| LDH X| LDH-C| LDH-X| LDH3| ldhc| LDHC_HUMAN| LDHX| MGC111073
- Cancer/testis antigen 32|CT32|EC 1.1.1.27|L lactate dehydrogenase C chain|L-lactate dehydrogenase C chain|Lactate Dehydrogenase C|Lactate dehydrogenase c variant 1|Lactate dehydrogenase c variant 3|Lactate dehydrogenase c variant 4|Lactate dehydrogenase C4|LDH C|LDH testis subunit|LDH X|LDH-C|LDH-X|LDH3|ldhc|LDHC_HUMAN|LDHX|MGC111073
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P07864
- P07864
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human L-lactate dehydrogenase C chain protein (2-332AA)
- Recombinant Human L-lactate dehydrogenase C chain protein (2-332AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice