Anti-KIF2A Rabbit Recombinant polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
0
Type:
Antigen: KIF2A
Clonality: Recombinant polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-582
- 77741-664
- Antigen Symbol:
- KIF2A
- KIF2A
- Antigen Synonyms:
- CDCBM3|HK2|KIF2|KIF2A_HUMAN|Kinesin 2|Kinesin heavy chain 2|Kinesin heavy chain member 2|Kinesin heavy chain member 2A |Kinesin like protein Kinesin-2|Kinesin-like protein KNS2|M Kinesin
- CDCBM3| HK2| KIF2| KIF2A_HUMAN| Kinesin 2| Kinesin heavy chain 2| Kinesin heavy chain member 2| Kinesin heavy chain member 2A | Kinesin like protein Kinesin-2| Kinesin-like protein KNS2| M Kinesin
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00139
- O00139
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Kinesin-like protein KIF2A protein (5-308AA)
- Recombinant Human Kinesin-like protein KIF2A protein (5-308AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice