Anti-MYSM1 Rabbit Polyclonal Antibody
77743-810
New Product
- Antigen Symbol:MYSM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q5VVJ2
- Antigen Synonyms:2A-DUB|2ADUB|Histone H2A deubiquitinase|Histone H2A deubiquitinase MYSM1|KIAA1915|Myb like SWIRM and MPN domains 1|Myb like, SWIRM and MPN domain containing protein|Myb like, SWIRM and MPN domains 1|Myb-like|Myb-like, SWIRM and MPN domains 1|MYSM 1|mysm1|MYSM1 Myb-like, SWIRM and MPN domains 1|MYSM1_HUMAN|RP4-592A1.1|SWIRM and MPN domain-containing protein 1
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthetic peptide derived from MYSM1
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: MYSM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human