Anti-KIF5B Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: KIF5B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77741-675
- 77858-590
- Antigen Symbol:
- KIF5B
- Antigen Synonyms:
- Conventional kinesin heavy chain|KIF 5B|KIF5B|Kinesin 1|kinesin 1 (110-120kD)|Kinesin 1 heavy chain|Kinesin family member 5B|Kinesin heavy chain|kinesin|heavy chain|ubiquitous|Kinesin-1 heavy chain|Kinesin1|KINH|KINH_HUMAN|KNS 1|KNS|KNS1|Ubiquitous kinesin heavy chain|UKHC
- Conventional kinesin heavy chain|KIF 5B|KIF5B|Kinesin 1|kinesin 1 (110-120kD)|Kinesin 1 heavy chain|Kinesin family member 5B|Kinesin heavy chain|kinesin|heavy chain|ubiquitous|Kinesin-1 heavy chain|Kinesin1|KINH|KINH_HUMAN|KNS 1|KNS|KNS1|Ubiquitous kinesin heavy chain|UKHC
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- P33176
- P33176
- Isotype:
- IgG
- Western Blot:
- Yes
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant Human Kinesin-1 heavy chain protein (338-514AA)
- Recombinant Human Kinesin-1 heavy chain protein (338-514AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice