Anti-HTR1A Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: HTR1A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77740-507
- 77857-739
- Antigen Symbol:
- HTR1A
- HTR1A
- Antigen Synonyms:
- 5 HT 1A|5 HT receptor 1A|5 HT1A|5 HT1A receptor|5 hydroxytryptamine (serotonin) receptor 1A|5 hydroxytryptamine (serotonin) receptor 1A G protein coupled|5 hydroxytryptamine receptor 1A|5-HT-1A|5-HT1A|5-hydroxytryptamine receptor 1A|5HT 1A|5HT 1A receptor|5HT receptor 1A|5HT1a|5HT1A_HUMAN|ADRB2RL1|ADRBRL1|G 21|G protein coupled receptor|G-21|Gpcr 18|Gpcr18|Guanine nucleotide binding regulatory protein-coupled receptor|PFMCD|Serotonin 1A receptor|Serotonin 5HT1A receptor|Serotonin receptor 1A
- 5 HT 1A|5 HT receptor 1A|5 HT1A|5 HT1A receptor|5 hydroxytryptamine (serotonin) receptor 1A|5 hydroxytryptamine (serotonin) receptor 1A G protein coupled|5 hydroxytryptamine receptor 1A|5-HT-1A|5-HT1A|5-hydroxytryptamine receptor 1A|5HT 1A|5HT 1A receptor|5HT receptor 1A|5HT1a|5HT1A_HUMAN|ADRB2RL1|ADRBRL1|G 21|G protein coupled receptor|G-21|Gpcr 18|Gpcr18|Guanine nucleotide binding regulatory protein-coupled receptor|PFMCD|Serotonin 1A receptor|Serotonin 5HT1A receptor|Serotonin receptor 1A
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P08908
- P08908
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human 5-hydroxytryptamine receptor 1A protein (230-334AA)
- Recombinant Human 5-hydroxytryptamine receptor 1A protein (230-334AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice