Anti-MSX1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:200 to 1:500
Type:
Antigen: MSX1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77860-058
- 77743-551
- Antigen Symbol:
- MSX1
- MSX1
- Antigen Synonyms:
- AA675338|AI324650|Homeobox 7|Homeobox protein Hox-7|Homeobox protein MSX 1|Homeobox protein MSX-1|Homeobox protein Homeobox, msh like 1|Homeobox, msh-like 1|HOX 7|Hox 7.1|Hox-7|HOX7|Hox7.1|HYD 1|HYD1|msh (Drosophila) homeo box homolog 1 (formerly homeo box 7)|Msh|msh homeo box 1|msh homeo box homolog 1|Msh homeobox 1|Msh homeobox 1 like protein|Msh homeobox 1-like protein|msh homeobox homolog 1 (Drosophila)|msh homeobox homolog 1|MSH, Drosophila, Homolog of, 1|MSX 1|MSX1_HUMAN|Muscle segment homeobox|Muscle segment homeobox, Drosophila, Homolog of, 1|OFC5|OTTHUMP00000115387|STHAG1
- AA675338|AI324650|Homeobox 7|Homeobox protein Hox-7|Homeobox protein MSX 1|Homeobox protein MSX-1|Homeobox protein Homeobox, msh like 1|Homeobox, msh-like 1|HOX 7|Hox 7.1|Hox-7|HOX7|Hox7.1|HYD 1|HYD1|msh (Drosophila) homeo box homolog 1 (formerly homeo box 7)|Msh|msh homeo box 1|msh homeo box homolog 1|Msh homeobox 1|Msh homeobox 1 like protein|Msh homeobox 1-like protein|msh homeobox homolog 1 (Drosophila)|msh homeobox homolog 1|MSH, Drosophila, Homolog of, 1|MSX 1|MSX1_HUMAN|Muscle segment homeobox|Muscle segment homeobox, Drosophila, Homolog of, 1|OFC5|OTTHUMP00000115387|STHAG1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P28360
- P28360
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Homeobox protein MSX-1 protein (19-179AA)
- Recombinant Human Homeobox protein MSX-1 protein (19-179AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice