Anti-FGF9 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: FGF9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77855-378
- 77737-148
- Antigen Symbol:
- FGF9
- FGF9
- Antigen Synonyms:
- FGF 9|FGF-9|FGF9_HUMAN|Fibroblast growth factor 9|GAF (Glia-activafibroblast growth factor 9 (glia-activating factor)|GAF|Glia Activating Factor|Glia-activating factor|HBFG 9|HBFG9|HBGF-9|Heparin-binding growth factor 9|MGC119914|MGC119915|SYNS3
- FGF 9|FGF-9|FGF9_HUMAN|Fibroblast growth factor 9|GAF (Glia-activafibroblast growth factor 9 (glia-activating factor)|GAF|Glia Activating Factor|Glia-activating factor|HBFG 9|HBFG9|HBGF-9|Heparin-binding growth factor 9|MGC119914|MGC119915|SYNS3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P31371
- P31371
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Fibroblast growth factor 9 protein (1-208AA)
- Recombinant Human Fibroblast growth factor 9 protein (1-208AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice