Anti-LYPD3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LYPD3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-267
- 77742-503
- Antigen Symbol:
- LYPD3
- Antigen Synonyms:
- 2310061G07Rik|C4.4A|GPI-anchored metastasis-associated protein C4.4A homolog|GPI-anchored metastasis-associated protein homolog|Ly6/PLAUR domain-containing protein 3|Lypd3|LYPD3_HUMAN|Matrigel-induced gene C4 protein|MIG-C4|SHINC3
- 2310061G07Rik|C4.4A|GPI-anchored metastasis-associated protein C4.4A homolog|GPI-anchored metastasis-associated protein homolog|Ly6/PLAUR domain-containing protein 3|Lypd3|LYPD3_HUMAN|Matrigel-induced gene C4 protein|MIG-C4|SHINC3
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- O95274
- O95274
- Isotype:
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human Ly6/PLAUR domain-containing protein 3 protein (31-326AA)
- Recombinant Human Ly6/PLAUR domain-containing protein 3 protein (31-326AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice