Anti-LIG3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: LIG3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77742-164
- 77858-975
- Antigen Symbol:
- LIG3
- LIG3
- Antigen Synonyms:
- DNA ligase|Ligase II|OTTMUSP00000000875|DNLI3_HUMAN|Polydeoxyribonucleotide synthase [ATP] 3|RP23-316C11.6|DNA ligase 3|LIG2|ATP dependent|LIG 3|MGC78176|Ligase III DNA ATP dependent|Polydeoxyribonucleotide synthase [ATP]|DNA|D11Wsu78e|Polydeoxyribonucleotide synthase|DNA ligase III
- D11Wsu78e|DNA ligase|DNA ligase 3|DNA ligase III|DNLI3_HUMAN|LIG 3|LIG2|Ligase II|DNA|ATP dependent|Ligase III DNA ATP dependent|MGC78176|OTTMUSP00000000875|Polydeoxyribonucleotide synthase [ATP] 3|Polydeoxyribonucleotide synthase [ATP]|Polydeoxyribonucleotide synthase|RP23-316C11.6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P49916
- P49916
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human DNA ligase 3 protein (809-928AA)
- Recombinant Human DNA ligase 3 protein (809-928AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice