Anti-ZSCAN22 Rabbit Polyclonal Antibody
77738-324
New Product
- Antigen Symbol:ZSCAN22
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P10073
- Antigen Synonyms:HKR2|ZNF50|Zinc finger and SCAN domain-containing protein 22|Krueppel-related zinc finger protein 2|Protein HKR2|Zinc finger protein 50
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Zinc finger protein 50
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA:1:10000
Type:
Antigen: ZSCAN22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human