Anti-OLIG1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: OLIG1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77745-454
- 77745-453
- Antigen Symbol:
- OLIG1
- OLIG1
- Antigen Synonyms:
- Basic domain helix loop helix protein class B 6|Basic domain helix loop helix protein class B6|BHLH B6|BHLHB 6|bHLHb6|bHLHe21|Class B basic helix-loop-helix protein 6|Class E basic helix-loop-helix protein 21|Olig 1|Olig1|OLIG1_HUMAN|Oligo 1|Oligo1|Oligodendrocyte lineage transcription factor 1|Oligodendrocyte specific bHLH transcription factor 1|Oligodendrocyte transcription factor 1
- Basic domain helix loop helix protein class B 6|Basic domain helix loop helix protein class B6|BHLH B6|BHLHB 6|bHLHb6|bHLHe21|Class B basic helix-loop-helix protein 6|Class E basic helix-loop-helix protein 21|Olig 1|Olig1|OLIG1_HUMAN|Oligo 1|Oligo1|Oligodendrocyte lineage transcription factor 1|Oligodendrocyte specific bHLH transcription factor 1|Oligodendrocyte transcription factor 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TAK6
- Q8TAK6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Oligodendrocyte transcription factor 1 protein (17-105AA)
- Oligodendrocyte transcription factor 1 protein (17-105AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice