Anti-HSF2 Rabbit Polyclonal Antibody
77740-446
New Product
- Antigen Symbol:HSF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Form:Liquid
- Gene ID:Q03933
- Antigen Synonyms:Heat shock factor protein 2|Heat shock transcription factor 2|HSF 2|HSF2|HSF2_HUMAN|HSTF 2
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:HSTF 2
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: HSF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat