Anti-HABP2 Rabbit Polyclonal Antibody
Catalog # 77739-394
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HABP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q14520
- Antigen Synonyms:Factor seven activating protease|Factor seven-activating protease|Factor VII activating protein|Factor VII-activating protease|FSAP|HABP 2|HABP|Habp2|HABP2_HUMAN|Hepatocyte growth factor activator like protein|Hepatocyte growth factor activator-like protein|HGFAL|Hyaluronan binding protein 2|Hyaluronan-binding protein 2 27 kDa light chain alternate form|Hyaluronic acid binding protein 2|PHBP|Plasma hyaluronan binding protein|Plasma hyaluronan-binding protein
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Plasma hyaluronan-binding protein
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: HABP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat