Anti-MARCKSL1 Rabbit Polyclonal Antibody
Catalog # 77742-783
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MARCKSL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P49006
- Antigen Synonyms:F52|Mac MARCKS|Mac-MARCKS|MacMARCKS|Macrophage enriched Myristoylated Alanine Rich C Kinase Substrate Like Protein|Macrophage myristoylated alanine-rich C kinase substrate|MARCKS like 1|MARCKS like protein 1|MARCKS related protein|MARCKS-like protein 1|MARCKS-related protein|MLP|MLP1|MRP|MRP_HUMAN
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:MRP_HUMAN
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MARCKSL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse