Anti-FHL1 Rabbit Polyclonal Antibody
Catalog # 77737-200
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:FHL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q13642
- Antigen Synonyms:bA535K18.1|FHL 1|FHL 1B|FHL-1|FHL1 protein|FHL1_HUMAN|FHL1A|FHL1B|FLH1A|Four and a half LIM domains 1|Four and a half LIM domains protein 1|Four and a half Lin11 Isl 1 and Mec 3 domains 1|four-and-a-half Lin11|Isl-1 and Mec-3|KYO T|KYOT|mouse|homolog of|LIM protein SLIMMER|MGC111107|RAM14-1|RBMX1A|RBMX1B|RBP associated molecule 14-1|RP11-535K18.1|Skeletal muscle LIM protein 1|Skeletal muscle LIM-protein 1|SLIM 1|SLIM|SLIM-1|SLIM1|SLIMMER|XMPMA
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:XMPMA
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: FHL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse