Anti-LDOC1 Rabbit Polyclonal Antibody
Catalog # 77736-854
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:LDOC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:50 µg
- Form:Liquid
- Gene ID:O95751
- Antigen Synonyms:BCUR1|Breast cancer up regulated 1|Breast cancer upregulated 1|Gm366|LDOC1_HUMAN|Leucine zipper down regulated in cancer 1|Leucine zipper protein down regulated in cancer cells|Leucine zipper protein down-regulated in cancer cells|Mar7|Mart7|Protein RGD1565644
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:BCUR1
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:5000
Type:
Antigen: LDOC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse