Anti-MAF Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MAF
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-292
- 77742-544
- Antigen Symbol:
- MAF
- MAF
- Antigen Synonyms:
- AS42 oncogene homolog|Avian musculoaponeurotic fibrosarcoma (MAF) protooncogene|Avian musculoaponeurotic fibrosarcoma (v maf)|c maf proto oncogene|cMAF_HUMAN|MAF2|MGC71685|Proto oncogene c Maf|Proto-oncogene c-Transcription factor v maf musculoaponeurotic fibrosarcoma oncogene homolog (avian)|v maf musculoaponeurotic fibrosarcoma oncogene homolog|V-maf musculoaponeurotic fibrosarcoma oncogene homolog
- AS42 oncogene homolog|Avian musculoaponeurotic fibrosarcoma (MAF) protooncogene|Avian musculoaponeurotic fibrosarcoma (v maf)|c maf proto oncogene|cMAF_HUMAN|MAF2|MGC71685|Proto oncogene c Maf|Proto-oncogene c-Transcription factor v maf musculoaponeurotic fibrosarcoma oncogene homolog (avian)|v maf musculoaponeurotic fibrosarcoma oncogene homolog|V-maf musculoaponeurotic fibrosarcoma oncogene homolog
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75444
- O75444
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Transcription factor Maf protein (55-180AA)
- Recombinant Human Transcription factor Maf protein (55-180AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice