Anti-IGFBP2 Rabbit Polyclonal Antibody
77740-756
New Product
- Antigen Symbol:IGFBP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P18065
- Antigen Synonyms:BP 2|BP2|IBP 2|IBP-2|IBP2|IBP2_HUMAN|IGF binding protein 2|IGF BP53|IGF-binding protein 2|IGFBP 2|IGFBP-2|IGFBP53|Insulin like growth factor binding protein 2 36kDa|Insulin like growth factor binding protein 2|Insulin like growth factor-binding protein 2 precursor|Insulin-like growth factor-binding protein 2
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Insulin-like growth factor-binding protein 2
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human