Anti-NUP98 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: NUP98
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77745-238
- 77745-237
- Antigen Symbol:
- NUP98
- NUP98
- Antigen Synonyms:
- 96 kDa nucleoporin|98 kDa nucleoporin|ADAR2|ADIR2|GLFG-repeat containing nucleoporin|Nuclear pore complex protein Nup96|Nuclear pore complex protein Nup98 Nup96|Nucleoporin 98kD|nucleoporin 98kDa|Nucleoporin Nup96|Nucleoporin NUP196|NUP96|Nup98-Nup96|NUP98_HUMAN
- 96 kDa nucleoporin|98 kDa nucleoporin|ADAR2|ADIR2|GLFG-repeat containing nucleoporin|Nuclear pore complex protein Nup96|Nuclear pore complex protein Nup98 Nup96|Nucleoporin 98kD|nucleoporin 98kDa|Nucleoporin Nup96|Nucleoporin NUP196|NUP96|Nup98-Nup96|NUP98_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P52948
- P52948
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Nuclear pore complex protein Nup98-Nup96 protein (965-1110AA)
- Nuclear pore complex protein Nup98-Nup96 protein (965-1110AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice