Anti-HNRNPH1 Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
0
Type:
Antigen: HNRNPH1
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77857-532
- 77740-243
- Antigen Symbol:
- HNRNPH1
- HNRNPH1
- Antigen Synonyms:
- DKFZp686A15170|Heterogeneous nuclear ribonucleoprotein H|Heterogeneous nuclear ribonucleoprotein H1 (H)|Heterogeneous nuclear ribonucleoprotein H1|HNRH1_HUMAN|hnRNP H|hnRNPH|HNRPH 1|HNRPH|HNRPH1 protein|N-terminally processed
- DKFZp686A15170| Heterogeneous nuclear ribonucleoprotein H| Heterogeneous nuclear ribonucleoprotein H1 (H)| Heterogeneous nuclear ribonucleoprotein H1| HNRH1_HUMAN| hnRNP H| hnRNPH| HNRPH 1| HNRPH| HNRPH1 protein| N-terminally processed
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P31943
- P31943
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Heterogeneous nuclear ribonucleoprotein H protein (2-449AA)
- Recombinant Human Heterogeneous nuclear ribonucleoprotein H protein (2-449AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice