Anti-MAT2B Rabbit Polyclonal Antibody
77742-818
New Product
- Antigen Symbol:MAT2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9NZL9
- Antigen Synonyms:1110064C04Rik| 2410018D16Rik| AI182287| AU022853| Beta regulatory subunit of methionine adenosyltransferase| dTDP 4 keto 6 deoxy D glucose 4 reductase| DTDP-4-keto-6-deoxy-D-glucose 4-reductase| MAT II| MAT II beta| MAT2B_HUMAN| MATIIbeta| Methionine adenosyltransferase 2 beta subunit| Methionine adenosyltransferase 2 subunit beta| Methionine adenosyltransferase II beta| MGC12237| MSTP045| Nbla02999| OTTMUSP00000005600| Putative dTDP-4-keto-6-deoxy-D-glucose 4-reductase| Putative protein product of Nbla02999| RP23-382C18.2| SDR23E1| Short chain dehydrogenase/reductase family 23E member 1| TGR
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:TGR
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: MAT2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat