Anti-GPR32 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GPR32
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-355
- 77738-856
- Antigen Symbol:
- GPR32
- Antigen Synonyms:
- G protein coupled receptor GPR32|GPR32|GPR32_HUMAN|Probable G protein coupled receptor 32|Probable G-protein coupled receptor 32
- G protein coupled receptor GPR32|GPR32|GPR32_HUMAN|Probable G protein coupled receptor 32|Probable G-protein coupled receptor 32
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- O75388
- O75388
- Isotype:
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human Probable G-protein coupled receptor 32 protein (1-44AA)
- Recombinant Human Probable G-protein coupled receptor 32 protein (1-44AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice