Anti-DDX3X Rabbit Polyclonal Antibody
Catalog # 77735-970
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:DDX3X
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:50 µg
- Form:Liquid
- Gene ID:O00571
- Antigen Synonyms:ATP dependent RNA helicase DDX3X|ATP-dependent RNA helicase DDX3X|CAP Rf|DBX|DDX14|DDX3X|DDX3X_HUMAN|DEAD (Asp Glu Ala Asp) box polypeptide 3 X linked|DEAD (Asp-Glu-Ala-Asp) box helicase 3|X-linked|DEAD box|DEAD box protein 3|DEAD box protein 3 X-chromosomal|DEAD box X isoform|X isoform|DEAD/H (Asp Glu Ala Asp/His) box polypeptide 3|DEAD/H box 3|Helicase like protein 2|Helicase-like protein 2|HLP2|X-chromosomal
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:ATP dependent RNA helicase DDX3X
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:100 to 1:300, IF: 1:200 to 1:1000, ELISA: 1:10000
Type:
Antigen: DDX3X
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat