Anti-NCOR2 Rabbit Polyclonal Antibody
77744-073
New Product
- Antigen Symbol:NCOR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Form:Liquid
- Gene ID:Q9Y618
- Antigen Synonyms:CTG 26|CTG repeat protein 26|CTG26|N CoR2|N-CoR2|NCoR 2|Ncor2|NCOR2_HUMAN|Nuclear Receptor Co-Repressor 2|Nuclear receptor corepressor 2|retinoic-acid-receptor-associated corepressor|Silencing Mediator for Retanoid and Thyroid Hormone Receptors|Silencing mediator of retinoic acid and thyroid hormone receptor|SMAP 270|SMAP270|SMRT|SMRTE|SMRTE tau|T3 receptor associating factor|T3 receptor-associating factor|Thyroid retinoic acid receptor associated corepressor|Thyroid-|TNRC 14|TNRC14|TRAC 1|TRAC|TRAC1
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:TRAC1
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: NCOR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat