Anti-MYT1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: MYT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77743-817
- 77743-816
- Antigen Symbol:
- MYT1
- MYT1
- Antigen Synonyms:
- C20orf36|KIAA0835|KIAA1050|MTF1|Myelin transcription factor 1|Myelin transcription factor I|MYT1_HUMAN|MyTI|PLPB1|Proteolipid protein-binding protein
- C20orf36|KIAA0835|KIAA1050|MTF1|Myelin transcription factor 1|Myelin transcription factor I|MYT1_HUMAN|MyTI|PLPB1|Proteolipid protein-binding protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q01538
- Q01538
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Myelin transcription factor 1 protein (317-451AA)
- Myelin transcription factor 1 protein (317-451AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice