Anti-FSHR Rabbit Polyclonal Antibody
Catalog # 77736-932
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:FSHR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P23945
- Antigen Synonyms:Follicle stimulating hormone receptor|Follicle stimulating hormone receptor isoform 1|Follicle-stimulating hormone receptor|Follitropin receptor|FSH receptor|FSH-R|FSHR_HUMAN|FSHRO|LGR1|MGC141667|MGC141668|ODG1|ovarian dysgenesis 1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Follicle stimulating hormone receptor
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IF: 1:200 to 1:1000, ELISA: 1:5000
Type:
Antigen: FSHR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human