Anti-INTS3 Rabbit Polyclonal Antibody
77741-126
New Product
- Antigen Symbol:INTS3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q68E01
- Antigen Synonyms:C1orf193|C1orf60|Integrator complex subunit 3|Int3|SOSS complex subunit A|Sensor of single-strand DNA complex subunit A|SOSS-A|Sensor of ssDNA subunit A
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Sensor of ssDNA subunit A
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: INTS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat