Anti-IL17RE Rabbit Polyclonal Antibody
Catalog # 77740-958
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:IL17RE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q8NFR9
- Antigen Synonyms:UNQ3056/PRO9877|Interleukin-17 receptor E|IL-17 receptor E|IL-17RE
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:IL-17RE
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: IL17RE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human