Anti-GSTA1 Rabbit Recombinant polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
0
Type:
Antigen: GSTA1
Clonality: Recombinant polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-527
- 77739-129
- Antigen Symbol:
- GSTA1
- GSTA1
- Antigen Synonyms:
- Glutathione S alkyltransferase A1|Glutathione S aryltransferase A1|Glutathione S transferase 2|Glutathione S transferase A1|Glutathione S transferase alpha 1|Glutathione S transferase Ha subunit 1|Glutathione S-transferase A1|GST class-alpha member 1|GST epsilon|GST HA subunit 1|GST|class alpha|1|GST-epsilon|GST2|GSTA1 1|GSTA1-1|GSTA1_HUMAN|GTH1|HA subunit 1|MGC131939|OTTHUMP00000016611|S (hydroxyalkyl)glutathione lyase A1
- Glutathione S alkyltransferase A1| Glutathione S aryltransferase A1| Glutathione S transferase 2| Glutathione S transferase A1| Glutathione S transferase alpha 1| Glutathione S transferase Ha subunit 1| Glutathione S-transferase A1| GST class-alpha member 1| GST epsilon| GST HA subunit 1| GST| class alpha| 1| GST-epsilon| GST2| GSTA1 1| GSTA1-1| GSTA1_HUMAN| GTH1| HA subunit 1| MGC131939| OTTHUMP00000016611| S (hydroxyalkyl)glutathione lyase A1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P08263
- P08263
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Glutathione S-transferase A1 protein (2-222AA)
- Recombinant Human Glutathione S-transferase A1 protein (2-222AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice