Anti-FADS1 Rabbit Recombinant polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
0
Type:
Antigen: FADS1
Clonality: Recombinant polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77736-425
- 77855-102
- Antigen Symbol:
- FADS1
- FADS1
- Antigen Synonyms:
- D5D| Delta 5 desaturase| Delta 5 fatty acid desaturase| Delta(5) desaturase| Delta(5) fatty acid desaturase| Delta-5 desaturase| FADS1_HUMAN| FADS6| FADSD5| Fatty acid desaturase 1| FLJ38956| Linoleoyl CoA desaturase (delta 6 desaturase) like 1| LLCDL1| TU12
- D5D|Delta 5 desaturase|Delta 5 fatty acid desaturase|Delta(5) desaturase|Delta(5) fatty acid desaturase|Delta-5 desaturase|FADS1_HUMAN|FADS6|FADSD5|Fatty acid desaturase 1|FLJ38956|Linoleoyl CoA desaturase (delta 6 desaturase) like 1|LLCDL1|TU12
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60427
- O60427
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Fatty acid desaturase 1 protein (1-121AA)
- Recombinant Human Fatty acid desaturase 1 protein (1-121AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice