Anti-MAP4K2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MAP4K2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-425
- 77742-705
- Antigen Symbol:
- MAP4K2
- MAP4K2
- Antigen Synonyms:
- B lymphocyte serine/threonine protein kinase|B lymphocyte serine/threonine-protein kinase|BL44|GC kinase|GCK|Germinal center kinase|germinal centre kinase (GC kinase)|M4K2_HUMAN|MAPK/ERK kinase kinase kinase 2|MEK kinase kinase 2|MEKKK 2|Mitogen activated protein kinase kinase kinase kinase 2|Mitogen-activated protein kinase kinase kinase kinase 2|Rab8 interacting protein|formerly|Rab8-interacting protein|RAB8IP
- Rab8-interacting protein|GC kinase|MEKKK 2|Mitogen activated protein kinase kinase kinase kinase 2|MEK kinase kinase 2|Germinal center kinase|germinal centre kinase (GC kinase)|RAB8IP|GCK|Mitogen-activated protein kinase kinase kinase kinase 2|B lymphocyte serine/threonine-protein kinase|MAPK/ERK kinase kinase kinase 2|M4K2_HUMAN|BL44|B lymphocyte serine/threonine protein kinase|formerly|Rab8 interacting protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q12851
- Q12851
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Mitogen-activated protein kinase kinase kinase kinase 2 protein (386-469AA)
- Recombinant Human Mitogen-activated protein kinase kinase kinase kinase 2 protein (386-469AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice