Anti-MX2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:200 to 1:500, IF: 1:50 to 1:200
Type:
Antigen: MX2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-669
- 77860-147
- Antigen Symbol:
- MX2
- MX2
- Antigen Synonyms:
- Interferon induced GTP binding protein Interferon regulated resistance GTP binding protein MXB|Interferon-induced GTP-binding protein Interferon-regulated resistance GTP-binding protein MxB|MX 2|MX dynamin like GTPase 2|MX2_HUMAN|MXB|Myxovirus (influenza virus) resistance 2|Myxovirus resistance 2|Myxovirus resistance protein 2|p78 related protein|p78-related protein|Second interferon induced protein p78
- Interferon induced GTP binding protein Interferon regulated resistance GTP binding protein MXB|Interferon-induced GTP-binding protein Interferon-regulated resistance GTP-binding protein MxB|MX 2|MX dynamin like GTPase 2|MX2_HUMAN|MXB|Myxovirus (influenza virus) resistance 2|Myxovirus resistance 2|Myxovirus resistance protein 2|p78 related protein|p78-related protein|Second interferon induced protein p78
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P20592
- P20592
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Interferon-induced GTP-binding protein Mx2 protein (548-688AA)
- Recombinant Human Interferon-induced GTP-binding protein Mx2 protein (548-688AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together