Anti-GSPT1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GSPT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-516
- 77739-118
- Antigen Symbol:
- GSPT1
- GSPT1
- Antigen Synonyms:
- ERF3AEukaryotic peptide chain release factor GTP-binding subunit ERF3A|Eukaryotic peptide chain release factor subunit 3a|eRF3a|G1 to S phase transition protein 1 homolog
- ERF3AEukaryotic peptide chain release factor GTP-binding subunit ERF3A|Eukaryotic peptide chain release factor subunit 3a|eRF3a|G1 to S phase transition protein 1 homolog
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P15170
- P15170
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Eukaryotic peptide chain release factor GTP-binding subunit ERF3A protein (6-142AA)
- Recombinant human Eukaryotic peptide chain release factor GTP-binding subunit ERF3A protein (6-142AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice