Anti-IRF9 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IRF9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-215
- 77741-211
- Antigen Symbol:
- IRF9
- Antigen Synonyms:
- IFN alpha responsive transcription factor subunit|IFN-alpha-responsive transcription factor subunit|Interferon regulatory factor 9|interferon stimulated transcription factor 3|Interferon-stimulated gene factor 3 gamma|interferon-stimulated transcription factor 3|gamma 48kDa|IRF 9|IRF-9|Irf9|IRF9_HUMAN|ISGF 3 gamma|ISGF-3 gamma|ISGF3|ISGF3 p48 subunit|ISGF3G|OTTHUMP00000164692|OTTHUMP00000164693|p48|Transcriptional regulator ISGF3 subunit gamma
- IFN alpha responsive transcription factor subunit|IFN-alpha-responsive transcription factor subunit|Interferon regulatory factor 9|interferon stimulated transcription factor 3|Interferon-stimulated gene factor 3 gamma|interferon-stimulated transcription factor 3|gamma 48kDa|IRF 9|IRF-9|Irf9|IRF9_HUMAN|ISGF 3 gamma|ISGF-3 gamma|ISGF3|ISGF3 p48 subunit|ISGF3G|OTTHUMP00000164692|OTTHUMP00000164693|p48|Transcriptional regulator ISGF3 subunit gamma
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- Q00978
- Q00978
- Isotype:
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human Interferon regulatory factor 9 protein (44-180AA)
- Recombinant Human Interferon regulatory factor 9 protein (44-180AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice