Anti-MED19 Rabbit Polyclonal Antibody
Catalog # 77742-953
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MED19
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Form:Liquid
- Gene ID:A0JLT2
- Antigen Synonyms:LCMR1|Lung cancer metastasis related protein 1|Lung cancer metastasis-related protein 1|med19|MED19_HUMAN|Mediator complex subunit 19|Mediator of RNA polymerase II transcription subunit 19
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Mediator of RNA polymerase II transcription subunit 19
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MED19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat