Anti-PSMB5 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PSMB5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77739-326
- 77856-651
- Antigen Symbol:
- PSMB5
- PSMB5
- Antigen Synonyms:
- DKFZp459C139|EC 3.4.25.1|LMPX|Macropain epsilon chain|MB1|MGC104214|MGC118075|MGC134464|Multicatalytic endopeptidase complex epsilon chain|Proteasome (prosome, macropain) subunit, beta type, 5|Proteasome beta 5 subunit|Proteasome catalytic subunit 3|Proteasome chain 6|Proteasome epsilon chain|Proteasome subunit beta type-5|Proteasome subunit MB1|Proteasome subunit X|Proteasome subunit, beta type, 5|Proteasome subunit, beta-5|PSB5_HUMAN|PSX large multifunctional protease X|X
- DKFZp459C139|EC 3.4.25.1|LMPX|Macropain epsilon chain|MB1|MGC104214|MGC118075|MGC134464|Multicatalytic endopeptidase complex epsilon chain|Proteasome (prosome, macropain) subunit, beta type, 5|Proteasome beta 5 subunit|Proteasome catalytic subunit 3|Proteasome chain 6|Proteasome epsilon chain|Proteasome subunit beta type-5|Proteasome subunit MB1|Proteasome subunit X|Proteasome subunit, beta type, 5|Proteasome subunit, beta-5|PSB5_HUMAN|PSX large multifunctional protease X|X
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P28074
- P28074
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Proteasome subunit beta type-5 protein (106-188AA)
- Recombinant human Proteasome subunit beta type-5 protein (106-188AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice