Anti-LOXL3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LOXL3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77742-293
- 77859-080
- Antigen Symbol:
- LOXL3
- LOXL3
- Antigen Synonyms:
- LOXL|Lysyl oxidase-like protein 3|Lysyl oxidase like 3|LOXL3_HUMAN|Lysyl oxidase homolog 3
- LOXL|LOXL3_HUMAN|Lysyl oxidase homolog 3|Lysyl oxidase like 3|Lysyl oxidase-like protein 3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P58215
- P58215
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Lysyl oxidase homolog 3 protein (401-608AA)
- Recombinant Human Lysyl oxidase homolog 3 protein (401-608AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice