Anti-RIDA Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RIDA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77740-390
- 77857-656
- Antigen Symbol:
- RIDA
- RIDA
- Antigen Synonyms:
- 14.5 kDa translational inhibitor protein|2-iminobutanoate/2-iminopropanoate deaminase|Heat responsive protein 12|Heat-responsive protein 12|Hrsp12|p14.5|Perchloric acid soluble protein|PSP|Reactive intermediate imine deaminase A homolog|Ribonuclease UK114|Translational inhibitor p14.5|Translational inhibitor protein p14.5|UK114|UK114 antigen homolog|UK114_HUMAN
- 14.5 kDa translational inhibitor protein|2-iminobutanoate/2-iminopropanoate deaminase|Heat responsive protein 12|Heat-responsive protein 12|Hrsp12|p14.5|Perchloric acid soluble protein|PSP|Reactive intermediate imine deaminase A homolog|Ribonuclease UK114|Translational inhibitor p14.5|Translational inhibitor protein p14.5|UK114|UK114 antigen homolog|UK114_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P52758
- P52758
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human 2-iminobutanoate/2-iminopropanoate deaminase protein (2-137AA)
- Recombinant human 2-iminobutanoate/2-iminopropanoate deaminase protein (2-137AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice