Anti-NR4A1 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: NR4A1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77744-978
- 77744-979
- Antigen Symbol:
- NR4A1
- NR4A1
- Antigen Synonyms:
- Early response protein NAK1|GFRP 1|GFRP|GFRP1|Growth factor inducible nuclear protein N10|Growth Factor Inducible Nuclear Protein NP10|Growth Factor Response Protein 1|Hbr1|HMR|Hormone Receptor|MGC9485|N10|N10 nuclear protein|NAK 1|NAK1|Nerve growth factor IB nuclear receptor variant 1|NGFIB|NP 10|NP10|NR4A1|NR4A1_HUMAN|Nuclear hormone receptor NUR/77|Nuclear Hormone Receptor TR3|Nuclear receptor subfamily 4 group A member 1|NUR77|mouse|homolog of|Orphan nuclear receptor HMR|Orphan nuclear receptor NR4A1|Orphan nuclear receptor TR3|Orphan receptor tr3|Receptor NGFIB|ST 59|ST-59|ST59|Steroid receptor TR3|Testicular receptor 3|TR 3|TR3|TR3 orphan receptor
- Early response protein NAK1|GFRP 1|GFRP|GFRP1|Growth factor inducible nuclear protein N10|Growth Factor Inducible Nuclear Protein NP10|Growth Factor Response Protein 1|Hbr1|HMR|Hormone Receptor|MGC9485|N10|N10 nuclear protein|NAK 1|NAK1|Nerve growth factor IB nuclear receptor variant 1|NGFIB|NP 10|NP10|NR4A1|NR4A1_HUMAN|Nuclear hormone receptor NUR/77|Nuclear Hormone Receptor TR3|Nuclear receptor subfamily 4 group A member 1|NUR77|mouse|homolog of|Orphan nuclear receptor HMR|Orphan nuclear receptor NR4A1|Orphan nuclear receptor TR3|Orphan receptor tr3|Receptor NGFIB|ST 59|ST-59|ST59|Steroid receptor TR3|Testicular receptor 3|TR 3|TR3|TR3 orphan receptor
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P22736
- P22736
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Nuclear receptor subfamily 4 group A member 1 protein (1-170AA)
- Nuclear receptor subfamily 4 group A member 1 protein (1-170AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice