Anti-CHD4 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CHD4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77857-794
- 77740-578
- Antigen Symbol:
- CHD4
- CHD4
- Antigen Synonyms:
- ATP dependent helicase CHD4|ATP-dependent helicase CHD4|CHD 4|CHD-4|CHD4|CHD4_HUMAN|Chromodomain helicase DNA binding protein 4|Chromodomain-helicase-DNA-binding protein 4|Mi 2 autoantigen 218 kDa protein|Mi 2b|Mi-2 autoantigen 218 kDa protein|Mi2 beta|Mi2-beta
- ATP dependent helicase CHD4|ATP-dependent helicase CHD4|CHD 4|CHD-4|CHD4|CHD4_HUMAN|Chromodomain helicase DNA binding protein 4|Chromodomain-helicase-DNA-binding protein 4|Mi 2 autoantigen 218 kDa protein|Mi 2b|Mi-2 autoantigen 218 kDa protein|Mi2 beta|Mi2-beta
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14839
- Q14839
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Chromodomain-helicase-DNA-binding protein 4 protein (1-219AA)
- Recombinant Human Chromodomain-helicase-DNA-binding protein 4 protein (1-219AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice