Anti-FKBP1A Rabbit Polyclonal Antibody
Catalog # 77737-228
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:FKBP1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P62942
- Antigen Synonyms:12 kDa FK506-binding protein| 12 kDa FKBP| Calstabin 1| FK506 binding protein 1| FK506 binding protein 12| FK506 binding protein 1A (12kD)| FK506 binding protein 1A 12kDa| FK506 binding protein 1A| FK506 binding protein T cell 12 kD| FK506 binding protein| T cell| 12 kD| FK506 binding protein12| FK506-binding protein 1A| FKB1A_HUMAN| FKBP 12| FKBP 1A| FKBP-12| FKBP-1A| FKBP1| FKBP12| FKBP12 Exip3| FKBP12C| Immunophilin FKBP12| Peptidyl prolyl cis trans isomerase| Peptidyl-prolyl cis-trans isomerase PKC12| PKCI2| PPIase| PPIase Protein kinase C inhibitor 2| Rotamase
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Rotamase
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: FKBP1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat